2-Bromo-5-nitrothiophene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15866-06
A15866-14
CAAAA15866-03EA
125.69
CAD
CAAAA15866-03
CAAAA15866-06
CAAAA15866-14
2-Bromo-5-nitrothiophene 97%
2-Bromo-5-nitrothiophene
Purity: 97%
CAS No.: 13195-50-1
Molecular Formula: C4H2BrNO2S
Formula Weight: 208.04
Melting Point: 42-46°
Boiling Point: 235-237°
MDL No.: MFCD00022493
Formula:
C₄H₂BrNO₂S MW: 208.04 g/mol Boiling Pt: 235…237 °C Melting Pt: 42…46 °C Storage Temperature: Ambient |
MDL Number:
MFCD00022493 CAS Number: 13195-50-1 EINECS: 236-155-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...