2,3,4-Trichlorobenzenesulfonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

A15865-03 A15865-06
CAAAA15865-03EA 59.25 CAD
CAAAA15865-03 CAAAA15865-06
2,3,4-Trichlorobenzenesulfonyl chloride 97%
2,3,4-Trichlorobenzenesulphonyl chloride

Purity: 97% CAS No.: 34732-09-7 Molecular Formula: C6H2Cl4O2S Formula Weight: 279.96 Melting Point: 61-65° Beilstein Reference: 2649436 UN No.: 3261 MDL No.: MFCD00024871

Formula: C₆H₂Cl₄O₂S
Melting Pt: 61…65 °C
Storage Temperature: Ambient
MDL Number: MFCD00024871
CAS Number: 34732-09-7
EINECS: 252-174-4
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR