3,5-Di-tert-butyl-4-hydroxybenzaldehyde 98+%

Supplier: Thermo Scientific Chemicals
Warning

A15864-06 A15864-14 A15864-22
CAAAA15864-06EA 52.14 CAD
CAAAA15864-06 CAAAA15864-14 CAAAA15864-22
3,5-Di-tert-butyl-4-hydroxybenzaldehyde 98+%
3,5-Di-tert-butyl-4-hydroxybenzaldehyde

Purity: 98+% CAS No.: 1620-98-0 Molecular Formula: C15H22O2 Formula Weight: 234.34 Melting Point: 187-189° Beilstein Reference: 982526

Formula: C₁₅H₂₂O₂
Boiling Pt: 332 °C (1013 hPa)
Melting Pt: 187…189 °C
Density: 1.16 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008826
CAS Number: 1620-98-0
EINECS: 216-592-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR