4-Quinolinol 98%

Supplier: Thermo Scientific Chemicals
Warning

A15859-09 A15859-03 A15859-06
CAAAAA15859-09EA 443.98 CAD
CAAAAA15859-09 CAAAA15859-03 CAAAA15859-06
4-Quinolinol 98%
4-Quinolinol

Purity: 98% CAS No.: 611-36-9 Molecular Formula: C9H7NO Formula Weight: 145.16 Melting Point: 201-205° Beilstein Reference: 1524969

Formula: C₉H₇NO
Melting Pt: 201…205 °C
Storage Temperature: Ambient
MDL Number: MFCD00006777
CAS Number: 611-36-9
EINECS: 210-268-2
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR