2-Bromoisobutyric acid 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Bromo-2-methylpropanoic acid, α-Bromoisobutyric acid, 2-Bromo-2-methylpropionic acid

A15854-14 A15854-22 A15854-36
CAAAA15854-14EA 51.67 CAD
CAAAA15854-14 CAAAA15854-22 CAAAA15854-36
2-Bromoisobutyric acid 98%
2-Bromoisobutyric acid

Purity: 98% CAS No.: 2052-01-9 Molecular Formula: C4H7BrO2 Formula Weight: 167.00 Flash Point: 110°(230°F) Melting Point: 42-46° Boiling Point: 198-200° Beilstein Reference: 1745543 Merck Reference: 14,1421 UN No.: 3261 MDL No.: MFCD00004168

Formula: C₄H₇BrO₂
Boiling Pt: 198…200 °C
Melting Pt: 42…46 °C
Density: 1.43 g/cm³ (20 °C)
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004168
CAS Number: 2052-01-9
EINECS: 218-139-2
UN: 3261
ADR: 8,III
Merck Index: 14,1421

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR