2-Chloro-6-fluoroaniline 98%

Supplier: Thermo Scientific Chemicals
Warning

A15848-03 A15848-06
CAAAA15848-03EA 70.47 CAD
CAAAA15848-03 CAAAA15848-06
2-Chloro-6-fluoroaniline 98%
2-Chloro-6-fluoroaniline

Purity: 98% CAS No.: 363-51-9 Molecular Formula: C6H5ClFN Formula Weight: 145.56 Boiling Point: 67-69°/14mm UN No.: 2810 MDL No.: MFCD00040309

Formula: C₆H₅ClFN
Boiling Pt: 67…69 °C/14 mm
Melting Pt: 31…32 °C
Density: 1.316
Storage Temperature: Ambient
MDL Number: MFCD00040309
CAS Number: 363-51-9
EINECS: 206-657-1
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR