4-Fluoro-2-nitrophenol 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15847-06
A15847-14
CAAAA15847-06EA
46.61
CAD
CAAAA15847-06
CAAAA15847-14
4-Fluoro-2-nitrophenol 98+%
4-Fluoro-2-nitrophenol
Purity: 98+%
CAS No.: 394-33-2
Molecular Formula: C6H4FNO3
Formula Weight: 157.10
Melting Point: 73-76°
Beilstein Reference: 1950412
Formula:
C₆H₄FNO₃ Melting Pt: 73…76 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042528 CAS Number: 394-33-2 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...