α-Bromo-3-chlorotoulene 97%

Supplier: Thermo Scientific Chemicals
Danger

A15844-09 A15844-18
CAAAA15844-09EA 49.93 CAD
CAAAA15844-09 CAAAA15844-18
α-Bromo-3-chlorotoulene 97%

Purity: 97% CAS No.: 766-80-3 Molecular Formula: C7H6BrCl Formula Weight: 205.49 Flash Point: 110°(230°F) Refractive Index: 1.5890 Odor: Pungent Boiling Point: 108-110°/10mm Beilstein Reference: 636504 UN No.: 3265 MDL No.: MFCD00000597

Formula: C₇H₆BrCl
Boiling Pt: 108…110 °C/10 mm
Melting Pt: 17.5 °C
Density: 1.567
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000597
CAS Number: 766-80-3
EINECS: 212-171-0
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR