4-Phenylbenzaldehyde 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Biphenyl-4-carbaldehyde, [1,1'-Biphenyl]-4-carbaldehyde, 4-Biphenylcarboxaldehyde, Biphenyl-4-carboxaldehyde, p-Biphenylylaldehyde, 4-Biphenylylaldehyde, 4-Biphenylcarbaldehyde

A15839-06 A15839-14 A15839-22
CAAAAA15839-06EA 59.09 CAD
CAAAAA15839-06 CAAAA15839-14 CAAAA15839-22
4-Phenylbenzaldehyde 98+%
4-Phenylbenzaldehyde

Purity: 98+% CAS No.: 3218-36-8 Molecular Formula: C13H10O Formula Weight: 182.22 Flash Point: 172°(341°F) Melting Point: 57-61° Boiling Point: 184°/11mm Beilstein Reference: 606693 MDL No.: MFCD00006947

Formula: C₁₃H₁₀O
Boiling Pt: 184 °C/11 mm
Melting Pt: 57…61 °C
Density: 1.107
Flash Pt: 172 °C(341 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006947
CAS Number: 3218-36-8
EINECS: 221-742-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR