(2-Chlorophenyl)acetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Chlorophenylacetic acid

A15834-09 A15834-18 A15834-30
CAAAA15834-09EA 50.09 CAD
CAAAA15834-09 CAAAA15834-18 CAAAA15834-30
(2-Chlorophenyl)acetic acid 98%
(2-Chlorophenyl)acetic acid

Purity: 98% CAS No.: 2444-36-2 Molecular Formula: C8H7ClO2 Formula Weight: 170.60 Odor: Odorless Melting Point: 94-97° Boiling Point: 145°/5mm Beilstein Reference: 1101523 MDL No.: MFCD00004317

Formula: (C₆H₄Cl)CH₂COOH
MW: 170.6 g/mol
Boiling Pt: 145 °C/5 mm
Melting Pt: 94…97 °C
Storage Temperature: Ambient
MDL Number: MFCD00004317
CAS Number: 2444-36-2
EINECS: 219-482-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR