tert-Butyl chloroacetate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A15833-14
A15833-22
A15833-36
CAAAA15833-14EA
44.4
CAD
CAAAA15833-14
CAAAA15833-22
CAAAA15833-36
tert-Butyl chloroacetate 98%
tert-Butyl chloroacetate
Purity: 98%
CAS No.: 107-59-5
Molecular Formula: C6H11ClO2
Formula Weight: 150.61
Flash Point: 46°(115°F)
Refractive Index: 1.4230
Odor: Ester-like
Boiling Point: 155°
Beilstein Reference: 1753006
Merck Reference: 14,1563
UN No.: 2929
MDL No.: MFCD00000930
Formula:
C₆H₁₁ClO₂ Boiling Pt: 155 °C Density: 1.053 Flash Pt: 46 °C (115 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000930 CAS Number: 107-59-5 EINECS: 203-506-1 UN: 2929 ADR: 6.1,II Merck Index: 14,1563 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...