3-Amino-2-chloropyridine 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15830-09
A15830-18
A15830-30
CAAAA15830-09EA
55.3
CAD
CAAAA15830-09
CAAAA15830-18
CAAAA15830-30
3-Amino-2-chloropyridine 98+%
3-Amino-2-chloropyridine
Purity: 98+%
CAS No.: 6298-19-7
Molecular Formula: C5H5ClN2
Formula Weight: 128.56
Flash Point: 185°(365°F)
Odor: Unpleasant
Boiling Point: 130-134°/22mm
Beilstein Reference: 109814
MDL No.: MFCD00006238
Formula:
C₅H₅ClN₂ Boiling Pt: 130…134 °C (22 mmHg) Melting Pt: 76…80 °C Flash Pt: 185 °C (365 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006238 CAS Number: 6298-19-7 EINECS: 228-572-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...