1,7-Octadiyne 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Octa-1,7-diyne
A15829-09
A15829-18
CAAAA15829-09EA
149.63
CAD
CAAAA15829-09
CAAAA15829-18
1,7-Octadiyne 98%
1,7-Octadiyne
Purity: 98%
CAS No.: 871-84-1
Molecular Formula: HC≡C(CH2)4C≡CH
Formula Weight: 106.17
Storage Temperature: 2°C to 8°C
Flash Point: 23°(73°F)
Refractive Index: 1.4460
Odor: Recognizable
Boiling Point: 135-136°
Beilstein Reference: 1697275
UN No.: 3295
MDL No.: MFCD00008580
Formula:
C₈H₁₀ MW: 106.17 g/mol Boiling Pt: 135…136 °C Melting Pt: –32 °C Density: 0.810 Flash Pt: 23 °C (73 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00008580 CAS Number: 871-84-1 EINECS: 212-815-0 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...