Choline chloride 98+%

Supplier: Thermo Scientific Chemicals

A15828-0E A15828-22 A15828-36
CAAAA15828-0EEA 360.24 CAD
CAAAA15828-0E CAAAA15828-22 CAAAA15828-36
Choline chloride 98+%
Choline chloride

An acyl group acceptor


Purity: 98+% CAS No.: 67-48-1 Molecular Formula: HOCH2CH2N(CH3)3Cl Formula Weight: 139.63 Physical Form: Solid Sensitivity: Hygroscopic Beilstein Reference: 3563126 Merck Reference: 14,2206 MDL No.: MFCD00011721


An acyl group acceptor

Formula: HOH₂CCH₂N(CH₃)₃Cl
Melting Pt: ∼ 305 °C (dec.)
Density: ~ 1.205 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011721
CAS Number: 67-48-1
EINECS: 200-655-4
Merck Index: 14,2206
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77b015e76f45084bc46da3249578a85652cd6802&vid=85c8a0a1b152b861b3bc3ec8fe25a70509221d85&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR