Sulfanilic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Aniline-4-sulphonic acid, 4-Aminobenzenesulphonic acid, 4-Aminobenzenesulfonic acid, Aniline-4-sulfonic acid

A15825-0B A15825-0I A15825-30
CAAAA15825-0BEA 127.51 CAD
CAAAA15825-0B CAAAA15825-0I CAAAA15825-30
Sulfanilic acid 98+%
Sulphanilic acid

Reagent for the detection of histidine; intermediate for dyes and pharmaceuticals


Purity: 98+% CAS No.: 121-57-3 Molecular Formula: C6H7NO3S Formula Weight: 173.19 Physical Form: Cryst powder Density: 1.485 Appearance: White to pale brown Melting Point: ca 365° Beilstein Reference: 908765 Merck Reference: 14,8926 MDL No.: MFCD00007886


Reagent for the detection of histidine; intermediate for dyes and pharmaceuticals

Formula: 4-(H₂N)C₆H₄SO₃H
MW: 173.19 g/mol
Boiling Pt: 363
Melting Pt: 279
Density: 1.485
Storage Temperature: Ambient
MDL Number: MFCD00007886
CAS Number: 121-57-3
EINECS: 204-482-5
Merck Index: 14,8926
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e032bed891f24ee48419dc3c7237b0293fd3e629&vid=28238ab814bb2b8d6eb5012387d8bf41528469e7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR