(±)-Epichlorhydrin 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (±)-1-Chloro-2,3-epoxypropane, Epichlorohydrin, (±)-3-Chloro-1,2-epoxypropane, 1-Chloro-2,3-epoxypropane, Epichlorhydrin, (±)-Glycerol epichlorhydrin, 3-Chloro-1,2-epoxypropane

A15823-0E A15823-22 A15823-36
CAAAA15823-0EEA 183.28 CAD
CAAAA15823-0E CAAAA15823-22 CAAAA15823-36
(±)-Epichlorhydrin 99%
(±)-Epichlorhydrin

A cyclodextrin cross-linking reagent


Purity: 99% CAS No.: 106-89-8 Molecular Formula: C3H5ClO Formula Weight: 92.53 Flash Point: 32°(90°F) Physical Form: Liquid Density: 1.182 Refractive Index: 1.4380 Appearance: Colorless Odor: Like chlorine Melting Point: -57° Boiling Point: 115-117° Beilstein Reference: 79785 Merck Reference: 14,3611 UN No.: 2023 MDL No.: MFCD00005132


A cyclodextrin cross-linking reagent

Formula: C₃H₅ClO
MW: 92.52 g/mol
Boiling Pt: 115…117 °C
Melting Pt: –57 °C
Density: 1.182
Flash Pt: 32 °C (90 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005132
CAS Number: 106-89-8
EINECS: 203-439-8
UN: 2023
ADR: 6.1 II
Merck Index: 14,3611

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR