3-Nitrobenzonitrile 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: m-Nitrobenzonitrile

A15821-14 A15821-30 A15821-09
CAAAAA15821-14EA 103.17 CAD
CAAAAA15821-14 CAAAAA15821-30 CAAAA15821-09
3-Nitrobenzonitrile 98%
3-Nitrobenzonitrile
Formula: C₇H₄N₂O₂
MW: 148.12 g/mol
Boiling Pt: 165 °C/21 mm
Melting Pt: 114…117 °C
Density: 0.33 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007194
CAS Number: 619-24-9
EINECS: 210-587-7
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR