L-Alanine 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
(S)-(+)-Alanine, H-Ala-OH, L(+)-Alanine, Alanine, L-α-Alanine, A, H-L-Ala-OH, α-Alanine
A15804-14
A15804-22
A15804-36
CAAAA15804-14EA
64.34
CAD
CAAAA15804-14
CAAAA15804-22
CAAAA15804-36
L-Alanine 99%
L-Alanine
Purity: 99%
CAS No.: 56-41-7
Molecular Formula: C3H7NO2
Formula Weight: 89.09
Melting Point: ca 295° dec.
Beilstein Reference: 1720248
Merck Reference: 14,204
Optical Rotation: +14° (c=5 in 5N HCl)
MDL No.: MFCD00064410
Formula:
C₃H₇NO₂ MW: 89.09 g/mol Boiling Pt: 189 °C (1013 hPa) Melting Pt: ∼ 295 °C (dec.) Density: 1.432 Storage Temperature: Ambient |
MDL Number:
MFCD00064410 CAS Number: 56-41-7 EINECS: 200-273-8 Merck Index: 14,204 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...