4-Hydroxybenzonitrile 98+%

Supplier: Thermo Scientific Chemicals
Warning

A15803-18 A15803-30 A15803-22 A15803-36
CAAAAA15803-18EA 115.97 CAD
CAAAAA15803-18 CAAAAA15803-30 CAAAA15803-22 CAAAA15803-36
4-Hydroxybenzonitrile 98+%
4-Hydroxybenzonitrile

Purity: 98+% CAS No.: 767-00-0 Molecular Formula: C7H5NO Formula Weight: 119.12 Melting Point: 110-114° Beilstein Reference: 386130

Formula: C₇H₅NO
Boiling Pt: 148 °C (1 mmHg)
Melting Pt: 110…114 °C
Density: ∼1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002312
CAS Number: 767-00-0
EINECS: 212-175-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR