6-Hydroxynicotinic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
6-Oxo-1,6-dihydropyridine-3-carboxylic acid, 6-Hydroxy-3-pyridinecarboxylic acid, 6-Hydroxyniacin, 6-Hydroxypyridine-3-carboxylic acid
A15802-06
A15802-14
A15802-22
CAAAA15802-06EA
39.18
CAD
CAAAA15802-06
CAAAA15802-14
CAAAA15802-22
6-Hydroxynicotinic acid 98%
6-Hydroxynicotinic acid
Metabolite of nicotinamide. Stimulates adrenocortical secretion
Purity: 98%
CAS No.: 5006-66-6
Molecular Formula: C6H5NO3
Formula Weight: 139.11
Melting Point: 314-316°
Beilstein Reference: 472182
Formula:
C₆H₅NO₃ Melting Pt: 314…316 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006277 CAS Number: 5006-66-6 EINECS: 225-682-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...