Isophthalaldehyde 98%

Supplier: Thermo Scientific Chemicals

Synonyms: m-Phthalaldehyde, Isophthaldicarboxaldehyde, Isophthaldialdehyde

A15788-06 A15788-14 A15788-22
CAAAA15788-06EA 96.06 CAD
CAAAA15788-06 CAAAA15788-14 CAAAA15788-22
Isophthalaldehyde 98%
Isophthalaldehyde

Purity: 98% CAS No.: 626-19-7 Molecular Formula: C8H6O2 Formula Weight: 134.13 Melting Point: 85-91° Boiling Point: 136°/17mm MDL No.: MFCD00003372

Formula: C₈H₆O₂
MW: 134.13 g/mol
Boiling Pt: 136 °C/17 mm
Melting Pt: 85…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00003372
CAS Number: 626-19-7
EINECS: 210-935-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR