o-Tolylacetic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Tolylacetic acid, (2-Methylphenyl)acetic acid

A15783-06 A15783-14 A15783-22
CAAAA15783-06EA 43.61 CAD
CAAAA15783-06 CAAAA15783-14 CAAAA15783-22
o-Tolylacetic acid 99%
o-Tolylacetic acid

Purity: 99% CAS No.: 644-36-0 Molecular Formula: C9H10O2 Formula Weight: 150.18 Melting Point: 86-90° Beilstein Reference: 1936952

Formula: (C₆H₄CH₃)CH₂COOH
MW: 150.18 g/mol
Boiling Pt: 283
Melting Pt: 86…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00004328
CAS Number: 644-36-0
EINECS: 211-416-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR