Diethyl succinate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Succinic acid diethyl ester, 1,4-Butanedioic acid diethyl ester

A15777-22 A15777-0C A15777-36
CAAAAA15777-22EA 48.82 CAD
CAAAAA15777-22 CAAAA15777-0C CAAAA15777-36
Diethyl succinate 98%
Diethyl succinate

Purity: 98% CAS No.: 123-25-1 Molecular Formula: C8H14O4 Formula Weight: 174.20 Flash Point: 90°(194°F) Refractive Index: 1.4200 Melting Point: -21° Boiling Point: 216-218° Beilstein Reference: 907645 Merck Reference: 14,8869 MDL No.: MFCD00009208

Formula: C₈H₁₄O₄
MW: 174.2 g/mol
Boiling Pt: 216…218 °C
Melting Pt: –21 °C
Density: 1.043
Flash Pt: 90 °C (194 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009208
CAS Number: 123-25-1
EINECS: 204-612-0
Merck Index: 14,8869

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR