4-Cyanobenzophenone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Benzoylbenzonitrile
A15764-03
A15764-06
CAAAAA15764-14EA
1914.55
CAD
CAAAAA15764-14
CAAAA15764-03
CAAAA15764-06
4-Cyanobenzophenone 98%
4-Cyanobenzophenone
Purity: 98%
CAS No.: 1503-49-7
Molecular Formula: C14H9NO
Formula Weight: 207.23
Melting Point: 110-114°
Beilstein Reference: 641181
UN No.: 3439
MDL No.: MFCD00016382
Formula:
C₁₄H₉NO Boiling Pt: 363 °C (1013 hPa) Melting Pt: 110…114 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016382 CAS Number: 1503-49-7 EINECS: 216-126-6 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...