Phenol 99+%, detached crystals
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Phenylalcohol, Carbolic acid, Hydroxybenzene
A15760-22
A15760-36
CAAAA15760-22EA
50.5
CAD
CAAAA15760-22
CAAAA15760-36
CAAAA15760-61
Phenol 99+%, detached crystals
Phenol
Purity: 99+%
CAS No.: 108-95-2
Molecular Formula: C6H6O
Formula Weight: 94.11
Flash Point: 79°(175°F)
Odor: Sweet
Boiling Point: 181°
Beilstein Reference: 969616
Merck Reference: 14,7241
UN No.: 1671
MDL No.: MFCD00002143
Formula:
C₆H₅OH Boiling Pt: 181 °C Melting Pt: 40…43 °C Density: 1.071 Flash Pt: 79 °C (175 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002143 CAS Number: 108-95-2 EINECS: 203-632-7 UN: 1671 ADR: 6.1,II Merck Index: 14,7241 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...