2-Chloroaniline 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A15758-36
A15758-0C
A15758-0E
CAAAAA15758-36EA
71.42
CAD
CAAAAA15758-36
CAAAA15758-0C
CAAAA15758-0E
2-Chloroaniline 98+%
2-Chloroaniline
Purity: 98+%
CAS No.: 95-51-2
Molecular Formula: C6H6ClN
Formula Weight: 127.57
Flash Point: 97°(206°F)
Refractive Index: 1.5880
Odor: Characteristic
Melting Point: -3°
Boiling Point: 207-209°
Beilstein Reference: 606077
Merck Reference: 14,2118
UN No.: 2019
MDL No.: MFCD00007656
Formula:
C₆H₆ClN Boiling Pt: 207…209 °C Melting Pt: –3 °C Density: 1.213 Flash Pt: 97 °C (206 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007656 CAS Number: 95-51-2 EINECS: 202-426-4 UN: 2019 ADR: 6.1,II Merck Index: 14,2118 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...