Sodium chlorodifluoroacetate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Sodium 2-chloro-2,2-difluoroacetate, Chlorodifluoroacetic acid sodium salt

A15756-14 A15756-22 A15756-36
CAAAA15756-14EA 132.72 CAD
CAAAA15756-14 CAAAA15756-22 CAAAA15756-36
Sodium chlorodifluoroacetate 97%
Sodium chlorodifluoroacetate

Purity: 97% CAS No.: 1895-39-2 Molecular Formula: ClCF2CO2Na Formula Weight: 152.46 Melting Point: ca 195° dec. Beilstein Reference: 4162363

Formula: C₂ClF₂NaO₂
MW: 152.46 g/mol
Melting Pt: ∼ 195 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064771
CAS Number: 1895-39-2
EINECS: 217-586-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR