5-Methoxysalicylaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Hydroxy-m-anisaldehyde, 2-Hydroxy-5-methoxybenzaldehyde

A15753-18 A15753-22 A15753-06 A15753-14
CAAAAA15753-18EA 668.34 CAD
CAAAAA15753-18 CAAAAA15753-22 CAAAA15753-06 CAAAA15753-14
5-Methoxysalicylaldehyde 98%
5-Methoxysalicylaldehyde

Purity: 98% CAS No.: 672-13-9 Molecular Formula: C8H8O3 Formula Weight: 152.15 Flash Point: 110°(230°F) Refractive Index: 1.5780 Melting Point: 13-16° Boiling Point: 248-252° Beilstein Reference: 1100532 MDL No.: MFCD00003332

Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 248…252 °C
Melting Pt: 13…16 °C
Density: 1,221
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003332
CAS Number: 672-13-9
EINECS: 211-589-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR