Homopiperazine 98%

Supplier: Thermo Scientific Chemicals
Danger

A15751-14 A15751-22
CAAAAA15751-36EA 2534.3 CAD
CAAAAA15751-36 CAAAA15751-14 CAAAA15751-22
Homopiperazine 98%
Homopiperazine

Purity: 98% CAS No.: 505-66-8 Molecular Formula: C5H12N2 Formula Weight: 100.17 Flash Point: 64°(147°F) Odor: Amine-like Boiling Point: 168-170° Beilstein Reference: 102711 UN No.: 3259 MDL No.: MFCD00006933

Formula: C₅H₁₂N₂
Boiling Pt: 168…170 °C
Melting Pt: 38…44 °C
Density: 0.95
Flash Pt: 64 °C (147 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006933
CAS Number: 505-66-8
EINECS: 208-016-1
UN: 2923
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR