2-Ethylaminoethanol 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
N-Ethyl-2-aminoethanol, N-Ethyl-2-ethanolamin, 2-(Ethylamino)ethan-1-ol
A15744-0F
A15744-AP
CAAAA15744-0FEA
210.14
CAD
CAAAA15744-0F
CAAAA15744-AP
2-Ethylaminoethanol 98%
2-Ethylaminoethanol
Purity: 98%
CAS No.: 110-73-6
Molecular Formula: C4H11NO
Formula Weight: 89.14
Flash Point: 71°(160°F)
Refractive Index: 1.441
Melting Point: -9°
Boiling Point: 168-170°
Beilstein Reference: 635671
UN No.: 2922
MDL No.: MFCD00002841
Formula:
C₄H₁₁NO MW: 89.14 g/mol Boiling Pt: 168…170 °C Melting Pt: –9 °C Density: 0.914 Flash Pt: 71 °C (160 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002841 CAS Number: 110-73-6 EINECS: 203-797-5 UN: 1719 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...