4-tert-Butylbenzaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15740-06
A15740-22
CAAAA15740-06EA
69.05
CAD
CAAAA15740-06
CAAAA15740-22
4-tert-Butylbenzaldehyde 97%
4-tert-Butylbenzaldehyde
Purity: 97%
CAS No.: 939-97-9
Molecular Formula: C11H14O
Formula Weight: 162.23
Flash Point: 101°(213°F)
Refractive Index: 1.5285
Boiling Point: 244-246°
Beilstein Reference: 1906461
UN No.: 3082
MDL No.: MFCD00035742
Formula:
C₁₁H₁₄O Boiling Pt: 244…246 °C Density: 0.970 Flash Pt: 101 °C (213 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00035742 CAS Number: 939-97-9 EINECS: 213-367-9 UN: 3082 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...