L(+)-Arginine 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15738-22
A15738-36
CAAAA15738-0EEA
7709.03
CAD
CAAAA15738-0E
CAAAA15738-22
CAAAA15738-36
L(+)-Arginine 98+%
L(+)-Arginine
Purity: 98+%
CAS No.: 74-79-3
Molecular Formula: C6H14N4O2
Formula Weight: 174.20
Physical Form: Powder
Sensitivity: Air Sensitive
Appearance: White
Melting Point: 226° dec.
Beilstein Reference: 1725413
Merck Reference: 14,780
Optical Rotation: +26° (c=1.6 in 6N HCl)
MDL No.: MFCD00002635
Formula:
C₆H₁₄N₄O₂ Boiling Pt: 368 °C (1013 hPa) Melting Pt: 226 °C (dec.) Density: 1.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002635 CAS Number: 74-79-3 EINECS: 200-811-1 Merck Index: 14,780 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...