Octadecyltrichlorosilane 95% cont. 5-10% branched isomers
Supplier: Thermo Scientific Chemicals
|
Danger
|
A15732-0B
A15732-18
A15732-30
CAAAA15732-0BEA
793.16
CAD
CAAAA15732-0B
CAAAA15732-18
CAAAA15732-30
Octadecyltrichlorosilane 95% cont. 5-10% branched isomers
Octadecyltrichlorosilane
Purity: 95%
CAS No.: 112-04-9
Molecular Formula: C18H37Cl3Si
Formula Weight: 387.94
Flash Point: 189°(372°F)
Refractive Index: 1.4600
Odor: Pungent
Melting Point: ca 20°
Boiling Point: 160-162°/3mm
Beilstein Reference: 1778847
UN No.: 1800
MDL No.: MFCD00000484
Formula:
C₁₈H₃₇Cl₃Si Boiling Pt: 160…162 °C/3 mm Melting Pt: ∼ 20 °C Density: 0.984 Flash Pt: 189 °C (372 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000484 CAS Number: 112-04-9 EINECS: 203-930-7 UN: 1800 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...