2-Methyl-2-pentenal mixture E/Z 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Methylpent-2-enal
A15729-14
A15729-22
A15729-36
CAAAA15729-14EA
56.09
CAD
CAAAA15729-14
CAAAA15729-22
CAAAA15729-36
2-Methyl-2-pentenal mixture E/Z 97%
2-Methyl-2-pentenal
Purity: 97%
CAS No.: 623-36-9
Molecular Formula: C6H10O
Formula Weight: 98.15
Flash Point: 31°(87°F)
Refractive Index: 1.4495
Odor: Pleasant
Melting Point: -90°
Boiling Point: 134-136°
Beilstein Reference: 506124
UN No.: 1988
MDL No.: MFCD00006978
Formula:
C₆H₁₀O Boiling Pt: 134…136 °C Melting Pt: –90 °C Density: 0.860 Flash Pt: 31 °C (87 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006978 CAS Number: 623-36-9 EINECS: 210-789-5 UN: 1988 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...