Dextrin, precipitated by alcohol
Supplier: Thermo Scientific Chemicals
A15717-22
A15717-36
CAAAA15717-22EA
71.89
CAD
CAAAA15717-22
CAAAA15717-36
Dextrin, precipitated by alcohol
Dextrin
CAS No.: 9004-53-9
Merck Reference: 14,2953
MDL No.: MFCD00081554
Formula:
(C₆H₁₂O₆)n Density: 1.45 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00081554 CAS Number: 9004-53-9 EINECS: 232-675-4 Merck Index: 14,2953 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...