3-Thenoic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Thiophenecarboxylic acid, Thiophene-3-carboxylic acid

A15712-06 A15712-14 A15712-22
CAAAA15712-06EA 132.25 CAD
CAAAA15712-06 CAAAA15712-14 CAAAA15712-22
3-Thenoic acid 99%
3-Thenoic acid

Purity: 99% CAS No.: 88-13-1 Molecular Formula: C5H4O2S Formula Weight: 128.15 Melting Point: 137-141° Beilstein Reference: 1994 Merck Reference: 14,9279 MDL No.: MFCD00005467

Formula: C₅H₄O₂S
MW: 128.15 g/mol
Melting Pt: 137…141 °C
Storage Temperature: Ambient
MDL Number: MFCD00005467
CAS Number: 88-13-1
EINECS: 201-802-5
Merck Index: 14,9279

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR