(±)-α-Methylbenzylamine 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A15711-AE
A15711-AP
CAAAA15711-AEEA
56.56
CAD
CAAAA15711-AE
CAAAA15711-AP
(±)-α-Methylbenzylamine 98+%
(±)-α-Methylbenzylamine
Purity: 98+%
CAS No.: 618-36-0
Molecular Formula: C8H11N
Formula Weight: 121.18
Flash Point: 79°(174°F)
Refractive Index: 1.5270
Odor: Odorless
Boiling Point: 69-70°/10mm
Beilstein Reference: 636127
Merck Reference: 14,6026
UN No.: 2735
MDL No.: MFCD00008069
Formula:
C₆H₅CH(NH₂)CH₃ Boiling Pt: 69…70 °C/10 mm Melting Pt: –65 °C Density: 0.945 Flash Pt: 79 °C (174 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008069 CAS Number: 618-36-0 EINECS: 210-545-8 UN: 2735 ADR: 8,III Merck Index: 14,6026 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...