2-Butyne-1-ol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Butyn-1-ol, But-2-yn-1-ol, 1-Hydroxy-2-butyne

A15709-06 A15709-14
CAAAA15709-06EA 88.32 CAD
CAAAA15709-06 CAAAA15709-14
2-Butyne-1-ol 98%
2-Butyne-1-ol

Purity: 98% CAS No.: 764-01-2 Molecular Formula: CH3C≡CCH2OH Formula Weight: 70.09 Flash Point: 51°(123°F) Refractive Index: 1.4530 Odor: Characteristic Melting Point: -2° Boiling Point: 142-143° Beilstein Reference: 1733676 UN No.: 1987 MDL No.: MFCD00002914

Formula: C₄H₆O
MW: 70.09 g/mol
Boiling Pt: 142…143 °C
Melting Pt: –2 °C
Density: 0.938
Flash Pt: 51 °C (123 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002914
CAS Number: 764-01-2
EINECS: 212-113-4
UN: 1987
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR