2-Chlorobenzamide 98%

Supplier: Thermo Scientific Chemicals
Warning

A15706-14 A15706-22
CAAAA15706-14EA 50.56 CAD
CAAAA15706-14 CAAAA15706-22
2-Chlorobenzamide 98%
2-Chlorobenzamide

Purity: 98% CAS No.: 609-66-5 Molecular Formula: C7H6ClNO Formula Weight: 155.58 Melting Point: 140-144° Beilstein Reference: 508510

Formula: C₇H₆ClNO
Melting Pt: 140…144 °C
Density: 1.34 g/cm³ (18 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007973
CAS Number: 609-66-5
EINECS: 210-195-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR