3-Methyl-2-cyclohexen-1-one 98%

Supplier: Thermo Scientific Chemicals
Warning

A15704-09 A15704-18 A15704-14 A15704-22
CAAAAA15704-09EA 119.29 CAD
CAAAAA15704-09 CAAAAA15704-18 CAAAA15704-14 CAAAA15704-22
3-Methyl-2-cyclohexen-1-one 98%
3-Methyl-2-cyclohexen-1-one

Purity: 98% CAS No.: 1193-18-6 Molecular Formula: C7H10O Formula Weight: 110.15 Flash Point: 68°(154°F) Refractive Index: 1.4940 Odor: Pleasant Boiling Point: 199-200° Beilstein Reference: 1560601 MDL No.: MFCD00001581

Formula: C₇H₁₀O
Boiling Pt: 199…200 °C
Melting Pt: 198…199 °C
Density: 0.971
Flash Pt: 68 °C(154 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001581
CAS Number: 1193-18-6
EINECS: 214-769-7
UN: 1224
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR