p-Anisic acid 98+%
Supplier: Thermo Scientific Chemicals
A15697-0I
A15697-0B
A15697-30
CAAAAA15697-0IEA
1039.64
CAD
CAAAAA15697-0I
CAAAA15697-0B
CAAAA15697-30
p-Anisic acid 98+%
p-Anisic acid
Purity: 98+%
CAS No.: 100-09-4
Molecular Formula: C8H8O3
Formula Weight: 152.15
Flash Point: 185°(365°F)
Odor: Odorless
pH: 3-4
Melting Point: 182-185°
Boiling Point: 275-277°
Beilstein Reference: 508910
Merck Reference: 14,666
MDL No.: MFCD00002542
Formula:
C₈H₈O₃ Boiling Pt: 275…277 °C Melting Pt: 182…185 °C Density: 1.385 Flash Pt: 185 °C(365 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002542 CAS Number: 100-09-4 EINECS: 202-818-5 Merck Index: 14,666 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...