Methyl-o-toluate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl 2-methylbenzoate, o-Toluic acid methyl ester, 2-Toluic acid methyl ester, Methyl-2-methylbenzoate, 2-Methylbenzoic acid methyl ester
A15695-14
A15695-22
A15695-36
CAAAA15695-14EA
54.67
CAD
CAAAA15695-14
CAAAA15695-22
CAAAA15695-36
Methyl-o-toluate 99%
Methyl o-toluate
Purity: 99%
CAS No.: 89-71-4
Molecular Formula: C9H10O2
Formula Weight: 150.18
Flash Point: 82°(179°F)
Refractive Index: 1.5190
Boiling Point: 207-208°
Beilstein Reference: 2041846
MDL No.: MFCD00008428
Formula:
C₉H₁₀O₂ MW: 150.18 g/mol Boiling Pt: 207…208 °C Melting Pt: –50 °C Density: 1,072 Flash Pt: 82 °C(179 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008428 CAS Number: 89-71-4 EINECS: 201-932-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...