Ethyl palmitate 97%
Supplier: Thermo Scientific Chemicals
A15694-14
A15694-22
A15694-36
CAAAA15694-14EA
107.91
CAD
CAAAA15694-14
CAAAA15694-22
CAAAA15694-36
Ethyl palmitate 97%
Ethyl palmitate
Purity: 97%
CAS No.: 628-97-7
Molecular Formula: CH3(CH2)14CO2CH2CH3
Formula Weight: 284.48
Flash Point: 178°(352°F)
Refractive Index: 1.4395
Melting Point: 22-26°
Boiling Point: 192-193°/10mm
Beilstein Reference: 1782663
MDL No.: MFCD00008996
Formula:
C₁₈H₃₆O₂ Boiling Pt: 192…193 °C/10 mm Melting Pt: 22…26 °C Density: 0.857 Flash Pt: 178 °C(352 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008996 CAS Number: 628-97-7 EINECS: 211-064-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...