2-Nitrobenzenesulfonamide 98+%

Supplier: Thermo Scientific Chemicals
Warning

A15689-09 A15689-22 A15689-06 A15689-14
CAAAAA15689-09EA 151.68 CAD
CAAAAA15689-09 CAAAAA15689-22 CAAAA15689-06 CAAAA15689-14
2-Nitrobenzenesulfonamide 98+%
2-Nitrobenzenesulphonamide

Purity: 98+% CAS No.: 5455-59-4 Molecular Formula: C6H6N2O4S Formula Weight: 202.19 Melting Point: 188-194° Beilstein Reference: 2214215

Formula: C₆H₆N₂O₄S
MW: 202.19 g/mol
Melting Pt: 188…194 °C
Storage Temperature: Ambient
MDL Number: MFCD00009807
CAS Number: 5455-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR