p-Phenylenediamine 97%

Supplier: Thermo Scientific Chemicals
Danger

A15680-0B A15680-0I A15680-30
CAAAA15680-0BEA 194.34 CAD
CAAAA15680-0B CAAAA15680-0I CAAAA15680-30
p-Phenylenediamine 97%
p-Phenylenediamine

MDL: MFCD00007901 Beilstein: 742029 Soluble in alcohol, chloroform, ether


Purity: 97% CAS No.: 106-50-3 Molecular Formula: C6H8N2 Formula Weight: 108.14 Flash Point: 156°(312°F) Physical Form: Cryst solid Odor: Weak Melting Point: 138-142° Boiling Point: 267-271° Solubility: Soluble in alcohol, chloroform, ether Beilstein Reference: 742029 Merck Reference: 14,7285 UN No.: 1673 MDL No.: MFCD00007901

Formula: C₆H₈N₂
Boiling Pt: 267…271 °C
Melting Pt: 138…142 °C
Density: 1.251 g/cm³ (20 °C)
Flash Pt: 156 °C (312 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007901
CAS Number: 106-50-3
EINECS: 203-404-7
UN: 1673
ADR: 6.1,III
Merck Index: 14,7285

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR