Sinapic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3,5-Dimethoxy-4-hydroxycinnamic acid, 3',5'-Dimethoxy-4'-hydroxycinnamic acid, 4-Hydroxy-3,5-dimethoxycinnamic acid
A15676-03
A15676-06
A15676-14
CAAAA15676-03EA
60.51
CAD
CAAAA15676-03
CAAAA15676-06
CAAAA15676-14
Sinapic acid 98%
Sinapic acid
Purity: 98%
CAS No.: 530-59-6
Molecular Formula: C11H12O5
Formula Weight: 224.21
Melting Point: ca 195° dec.
Beilstein Reference: 2130006
Formula:
C₁₁H₁₂O₅ MW: 224.21 g/mol Boiling Pt: 378 °C (1013 hPa) Melting Pt: ∼ 195 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00004401 CAS Number: 530-59-6 EINECS: 208-487-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...