Hexylamine 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-Hexylamine, 1-Aminohexane
A15663-AK
A15663-AU
CAAAA15663-AKEA
117.87
CAD
CAAAA15663-AK
CAAAA15663-AU
Hexylamine 99%
Hexylamine
Purity: 99%
CAS No.: 111-26-2
Molecular Formula: C6H15N
Formula Weight: 101.19
Flash Point: 27°(80°F)
Refractive Index: 1.4180
Odor: Amine-like
pH: 11.6
Melting Point: -19°
Boiling Point: 129-131°
Beilstein Reference: 1731298
UN No.: 2734
MDL No.: MFCD00008240
Formula:
H₃C(CH₂)₄CH₂NH₂ Boiling Pt: 129…131 °C Melting Pt: –19 °C Density: 0.773 Flash Pt: 27 °C (80 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008240 CAS Number: 111-26-2 EINECS: 203-851-8 UN: 2734 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...