4-Hexylbenzoyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

A15659-22 A15659-14
CAAAAA15659-22EA 922.72 CAD
CAAAAA15659-22 CAAAA15659-14
4-Hexylbenzoyl chloride 98%
4-Hexylbenzoyl chloride

Purity: 98% CAS No.: 50606-95-6 Molecular Formula: C13H17ClO Formula Weight: 224.73 Flash Point: 110°(230°F) Refractive Index: 1.5256 Boiling Point: 176-177°/22mm Beilstein Reference: 2208938 UN No.: 3265 MDL No.: MFCD00000701

Formula: C₁₃H₁₇ClO
Boiling Pt: 176…177 °C/22 mm
Density: 1.029
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000701
CAS Number: 50606-95-6
EINECS: 256-647-6
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR