Methyl decanoate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15658-22
A15658-18
A15658-30
CAAAAA15658-22EA
122.45
CAD
CAAAAA15658-22
CAAAA15658-18
CAAAA15658-30
Methyl decanoate 99%
Methyl decanoate
Purity: 99%
CAS No.: 110-42-9
Molecular Formula: CH3(CH2)8CO2CH3
Formula Weight: 186.30
Flash Point: 94°(201°F)
Refractive Index: 1.4250
Melting Point: -18°
Boiling Point: 223-225°
Beilstein Reference: 1759170
MDL No.: MFCD00009580
Formula:
C₁₁H₂₂O₂ Boiling Pt: 223…225 °C Melting Pt: –18 °C Density: 0.877 Flash Pt: 94 °C(201 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009580 CAS Number: 110-42-9 EINECS: 203-766-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...