4-Chlorophenylboronic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

A15657-03 A15657-06 A15657-14
CAAAA15657-03EA 39.18 CAD
CAAAA15657-03 CAAAA15657-06 CAAAA15657-14
4-Chlorophenylboronic acid 98+%
4-Chlorophenylboronic acid

Purity: 98+% CAS No.: 1679-18-1 Molecular Formula: C6H6BClO2 Formula Weight: 156.38 Melting Point: 268-277° Beilstein Reference: 2936346

Formula: C₆H₆BClO₂
Melting Pt: 268…277 °C
Density: ~ 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00039137
CAS Number: 1679-18-1
EINECS: 216-845-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR